Learn More
FSI North AmericaSupplier Diversity Partner FSI DAT Series Pneumatic Shelter DAT3015 DFS Item
Manufacturer:  FSI North AmericaSupplier Diversity Partner DAT3015
Approx. 10’ W X 15’ L X 9’ H (3 M X 4.5 M X 2.7 M) – Shelter only, 150 sq. ft. (13.5 M2) 200 lbs. (90 kg) Carton size approx. = 136 X 108 X 91 cm (54” L X 32” W X 36” H).  2 windows – one on each side, 2 skylights, 4 (four) 17”/43 cm HVAC/clothes inlet/outlets with tie closures – 2 each on opposite ends/sides, 10 metal stakes and 4 tie down ropes.  Air volume approx. 90 cu. Ft; approx. 70 mph wind resistance; approx. roof load 1,000 lbs/8 lbs/sq ft. Inflator/deflator included.
Catalog No. 50-195-2460