Learn More
FSIP1 Mouse anti-Human, Clone: OTI6F6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806088
Catalog No. 50-166-7691
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact functions of FSIP1 remain unknown.
Specifications
Specifications
FSIP1
Monoclonal
Unconjugated
FSIP1
Lyophilized
FSIP1
Full length human recombit protein of human FSIP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6F6
PBS with 8% trehalose and no preservative; pH 7.3
Q8NA03
HSD10
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
161835