Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559386
Description
Immunogen sequence: PVKGYELAVT QHQQLAEIDI KLQELSAASP TISSFSPRLE NRNNQKPDRD GERNMEVTPG EKILRNTKEQ RDLHNRLREI DEKLKMMKEN V Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 57%.
The exact functions of FSIP1 remain unknown.
Specifications
FSIP1 | |
Polyclonal | |
Unconjugated | |
FSIP1 | |
1700012M13Rik; 4933432K11Rik; fibrous sheath interacting protein 1; fibrous sheath-interacting protein 1; Fsip1; HSD10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
161835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NA03 | |
FSIP1 | |
Recombinant protein corresponding to Human FSIP1. Recombinant protein control fragment (Product #RP-98100). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction