Learn More
Invitrogen™ FSTL4 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA548073
Description
This antibody detects recombinant mouse Follistatin-like 4/FSTL4 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FSTL4 is observed, and less than 5% cross-reactivity with rhFSTL5 and recombinant mouse FSTL1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
FSTL4 (Follistatin Like 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includecalcium ion binding. An important paralog of this gene isFSTL5.
Specifications
FSTL4 | |
Polyclonal | |
Unconjugated | |
FSTL4 | |
B230374F23Rik; follistatin like 4; follistatin-like 4; follistatin-like protein 4; Follistatin-related protein 4; FSL4; FSTL4; KIAA1061; m-D/Bsp120I 1-1; mKIAA1061; SPIG1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23105, 303130, 320027 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q5STE3, Q6MZW2 | |
FSTL4 | |
CHO-derived recombinant mouse Follistatin-like 4/FSTL4 Trp23-Val841. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.