Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSTL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548593
Description
FSTL5 Polyclonal Antibody for Western Blot

Specifications
FSTL5 | |
Polyclonal | |
Unconjugated | |
FSTL5 | |
9130207J01Rik; follistatin like 5; follistatin-like 5; follistatin-like protein 5; Follistatin-related protein 5; Fstl5; Kiaa1263; m-D/Bsp120I 1-2; ollistatin-like 5 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
213262, 56884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8BFR2, Q8N475 | |
FSTL5 | |
KLH conjugated synthetic peptide between 20-48 amino acids from the N-terminal region of human FSTL5. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction