Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FTBMT
GREENER_CHOICE

Catalog No. p-7161625
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective GPR52 agonist; orally bioavailable and BBB permeable

FTBMT (also known as TP 024) is a potent and selective GPR52 agonist (EC50 = 75 nM, Emax = 122%), which is selective for GPR52 over a panel of 98 targets including D1, D2, AMPA and NMDA. FTBMT suppresses methamphetamine-induced hyperlocomotion in mice, and inhibits MK-801 -induced hyperactivity (model for acute psychosis), without causing catalepsy in mice. This compound is orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material 4-[3-[[3-Fluoro-5-(trifluoromethyl)phenyl]methyl]-5-methyl-1H-1,2,4-triazol-1-yl]-2-methylbenzamide
CAS 1358575-02-6
Quantity 25 mg
Molecular Formula C19H16F4N4O
Formula Weight 392.35
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.