Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTI 277, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Prodrug form of FTI 276 (Cat. No. 2406) that inhibits farnesyltransferase (FTase) (IC50 = 0.5 nM). Inhibits H-Ras and K-Ras processing in whole cells (IC50 values are 0.1 and 10 μM respectively) and disrupts constitutive H-Ras-specific activation of MAPK. Causes significant antiproliferative effects in human malignant glioma cells and many other tumor cell lines.
Specifications
Quantity | 1 mg |
Formula Weight | 561.64 |
Percent Purity | >95% |
Chemical Name or Material | FTI 277 |
Safety and Handling
Recommended Storage | Store at -80°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction