Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTO Mouse anti-Human, Rat, Clone: 5-2H10, Abnova™

Mouse monoclonal antibody raised against rat Fto.

Manufacturer:  Abnova Corporation MAB12718

Catalog No. 89-934-168


Add to cart

Description

Description

Specifications

Specifications

FTO
5-2H10
Mouse monoclonal antibody raised against rat Fto.
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
RGD1305121
Mouse
IgG
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Rat
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
Fto
A synthetic peptide corresponding to N-terminus region of rat Fto.
Protein G purification
RUO
Primary
291905
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit