Sino Biological™ FTO Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human FTO (Lys 2-Pro 505) fused to polyhistide tag at N-terminus

Manufacturer: Sino Biological™ 12125H07E25

 View more versions of this product

Catalog No. 12-125-H07E25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Source Human
    Common Name Fat mass and Obesity associated
    Gene Symbol FTO
    Protein Tag His Tag
    Expression System E.coli
    Product Type FTO Recombinant Human Protein
    Format Lyophilized
    Molecular Weight 59.5kd
    Validated Application Assay Standard
    Protein Family Methylation and Demethylation Proteins
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Gene ID (Entrez) 79608

    Expressed from DNA sequence encoding human FTO (Lys 2-Pro 505) fused to polyhistide tag at N-terminus

    • Formulation: 20mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit