Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ftorafur, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
17902-23-7
C8H9FN2O3
200.169
MFCD00012351
WFWLQNSHRPWKFK-UHFFFAOYSA-N
tegafur,ftorafur,futraful,fluorofur,sinoflurol,citofur,fental,neberk,coparogin,florafur
5386
CHEBI:32188
5-fluoro-1-(oxolan-2-yl)pyrimidine-2,4-dione
C1CC(OC1)N2C=C(C(=O)NC2=O)F
17902-23-7
C8H9FN2O3
200.169
MFCD00012351
WFWLQNSHRPWKFK-UHFFFAOYSA-N
tegafur,ftorafur,futraful,fluorofur,sinoflurol,citofur,fental,neberk,coparogin,florafur
5386
CHEBI:32188
5-fluoro-1-(oxolan-2-yl)pyrimidine-2,4-dione
C1CC(OC1)N2C=C(C(=O)NC2=O)F
Color White
Melting Point 168.0°C to 171.0°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 250mg
Assay Percent Range 99%
Merck Index 15, 9252
Formula Weight 200.17
Physical Form Very Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material Ftorafur, 99%
Show More Show Less

EINECSNumber : 241-846-2

RTECSNumber : YR0450000

TSCA : TSCA