Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FTSJD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570294
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap2 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the second nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) (cap0) to produce m(7)GpppRmpNm (cap2). Recognizes a guanosine cap on RNA independently of its N(7) methylation status. Display cap2 methylation on both cap0 and cap1. Displays a preference for cap1 RNAs.
Specifications
FTSJD1 | |
Polyclonal | |
Unconjugated | |
CMTR2 | |
adrift homolog; AFT; AU022703; C730036L12Rik; cap methyltransferase 2; cap2 2'O-ribose methyltransferase 2; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 2; Cmtr2; FtsJ methyltransferase domain containing 1; ftsJ methyltransferase domain-containing protein 1; ftsjd1; HMTr2; hypothetical protein LOC406742; MTr2; Protein adrift homolog; RGD1309394; wu:fj48c05; zgc:55686 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IYT2 | |
CMTR2 | |
synthetic peptide directed towards the middle region of human FTSJD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction