Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FUCA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570565
Description
This target displays homology in the following species: Cow: 92%; Dog: 77%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 93%; Rat: 100%; Zebrafish: 92%.
FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
Specifications
FUCA2 | |
Polyclonal | |
Unconjugated | |
FUCA2 | |
0610025O11Rik; 5530401P20Rik; Alpha-L-fucosidase 2; alpha-L-fucoside fucohydrolase 2; dJ20N2.5; fc20c06; Fuca2; fucosidase, alpha-L- 2, plasma; MGC1314; plasma alpha-L-fucosidase; PSEC0151; UNQ227/PRO260; wu:fc20c06; zgc:92013 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66848 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99KR8 | |
FUCA2 | |
synthetic peptide directed towards the middle region of mouse FUCA2. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction