Learn More
Fucose Mutarotase Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP300239AF647
Catalog No. NB065016
Add to Cart
Description
Description
Fucose Mutarotase Polyclonal specifically detects Fucose Mutarotase in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Fucose Mutarotase
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
C10orf125, chromosome 10 open reading frame 125, FucM, fucose mutarotase, FUCU
Produced in rabbits immunized with purified, recombinant Fucose Mutarotase (A2VDF0-1; Met1-Leu154).
0.1 mL
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Affinity purified
RUO
282969
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions