Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fucosyltransferase 2/FUT2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30364520UL

 View more versions of this product

Catalog No. NB094823


Add to cart

Description

Description

Fucosyltransferase 2/FUT2 Polyclonal antibody specifically detects Fucosyltransferase 2/FUT2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Fucosyltransferase 2/FUT2
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
PBS with 50% glycerol, pH7.3.
alpha (1,2) fucosyltransferase, Alpha(1,2)FT 2, alpha(1,2)FT2, B12QTL1, EC 2.4.1.69, Fucosyltransferase 2, fucosyltransferase 2 (secretor status included), galactoside 2-alpha-L-fucosyltransferase 2, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2, Se, SE2, SEC2SE, Secretor blood group alpha-2-fucosyltransferase, Secretor factor, sej
Recombinant fusion protein containing a sequence corresponding to amino acids 29-150 of human Fucosyltransferase 2/FUT2 (NP_001091107.1).
Affinity purified
RUO
Primary
2524
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit