Alfa Aesar™ Fullerene, buckytube/nanotube, multi-walled, ground core, 7-12nm OD, 0.5-10 micron long

Manufacturer: Alfa Aesar™ 4154903

Quantity 1g

 View more versions of this product

Catalog No. AA4154903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 308068-56-6
    MDL Number MFCD00282904
    UN Number UN1325
    Chemical Name or Material Fullerene, buckytube/nanotube, Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    Color Black
    MDL Number MFCD00282904
    CAS 308068-56-6
    Health Hazard 1 Warning
    Quantity 1g
    Mesh Size −100 Mesh
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    UN Number UN1325
    Chemical Name or Material Fullerene, buckytube/nanotube, Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit