Learn More
Alfa Aesar™ Fullerene, buckytube/nanotube, single walled, as-produced
Manufacturer:  Alfa Aesar™ 4154803
Quantity 1g
 View more versions of this product
Catalog No. AA4154803
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
308068-56-6
MFCD00282904
UN1325
Fullerene, buckytube/nanotube, Single walled, as-produced
Specifications
308068-56-6
MFCD00282904
Black
Fullerene, buckytube/nanotube, Single walled, as-produced
Powder
UN1325
1g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.