Alfa Aesar™ Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long

Manufacturer: Alfa Aesar™ 43839MD

Quantity 250mg

 View more versions of this product

Catalog No. AA43839MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 308068-56-6
    MDL Number MFCD00133992
    UN Number UN1325
    Synonym Buckminster Buckytubes; Buckytubes
    Chemical Name or Material Fullerene, Nanotube, multi-walled, 20nm OD, 5 to 20 micron long

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Buckminster Buckytubes; Buckytubes
    MDL Number MFCD00133992
    Health Hazard 1 Warning
    Color Black
    Quantity 250mg
    Physical Form Powder
    CAS 308068-56-6
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Mesh Size −270 Mesh
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    UN Number UN1325
    Chemical Name or Material Fullerene, Nanotube, multi-walled, 20nm OD, 5 to 20 micron long

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit