Learn More
Fullerene, nanotube, multi-walled, as-produced cathode deposits, core and shell, Thermo Scientific Chemicals
Description
Multi-walled carbon nanotube were used in the comparative study of oxidative treatment to carbon nonmaterial's. Used as a reinforcement in CNT/SiC nano composites. Also used as fiber optics transducers, fiber optic tips for micro bubble generation in liquids, battery cathodes and high strength composites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 2 g |
UN Number | UN1325 |
Solubility Information | Insoluble in water and organic solvents. |
Chemical Name or Material | Fullerene, Nanotube, multi-walled, as-produced cathode deposits, core and shell |
Safety and Handling
Hazard Category | H228-H315-H319-H335 |
Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.