Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaraldehyde bis(dimethylacetal), 90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6068-62-8
C8H16O4
176.21
MFCD00075507
ZFGVCDSFRAMNMT-AATRIKPKSA-N
fumaraldehyde bis dimethyl acetal,e-1,1,4,4-tetramethoxybut-2-ene,fumaraldehyde bis dimethylacetal,2-butenedial bis dimethyl acetal,1,1,4,4-tetramethoxy-trans-2-butene,trans-1,1,4,4-tetramethoxy-2-butene,1,1,4,4-tetramethoxy-but-2-ene
5862719
(E)-1,1,4,4-tetramethoxybut-2-ene
COC(C=CC(OC)OC)OC
6068-62-8
C8H16O4
176.21
MFCD00075507
ZFGVCDSFRAMNMT-AATRIKPKSA-N
fumaraldehyde bis dimethyl acetal,e-1,1,4,4-tetramethoxybut-2-ene,fumaraldehyde bis dimethylacetal,2-butenedial bis dimethyl acetal,1,1,4,4-tetramethoxy-trans-2-butene,trans-1,1,4,4-tetramethoxy-2-butene,1,1,4,4-tetramethoxy-but-2-ene
5862719
(E)-1,1,4,4-tetramethoxybut-2-ene
COC(C=CC(OC)OC)OC
Specific Gravity 1.01
Flash Point 80°C
Boiling Point 100°C to 103°C (15.0mmHg)
Density 1.0100g/mL
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Colorless to Yellow
Assay Percent Range 88% min. (GC)
Linear Formula (CH3O)2CHCH=CHCH(OCH3)2
Formula Weight 176.21
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Fumaraldehyde bis(dimethylacetal)
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning