Alfa Aesar™ Fumaramide, 97%

Manufacturer: Alfa Aesar™ B2520022

Quantity 100g

 View more versions of this product

Catalog No. AAB2520022

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 627-64-5
    Molecular Formula C4H6N2O2
    Formula Weight 114.11
    MDL Number MFCD00025527
    Beilstein 1721338
    Chemical Name or Material Fumaramide

    Specifications

    Formula Weight 114.11
    Molecular Formula C4H6N2O2
    MDL Number MFCD00025527
    Beilstein 1721338
    Melting Point ≈266°C (dec.)
    Quantity 100g
    CAS 627-64-5
    Chemical Name or Material Fumaramide
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit