Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumarase Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: 7612, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243578C

Catalog No. NBP243578C


Add to cart

Description

Description

Fumarase Monoclonal antibody specifically detects Fumarase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Fumarase
7612
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
FH
Recombinant protein encompassing a sequence within the center region of human Fumarate hydratase. The exact sequence is proprietary.
Protein G purified
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
2271
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1
Mouse
IgG3
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit