Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumarase Mouse anti-Human, Rat, Canine, PE/Cy5.5, Clone: OTI1F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270733PECY55

Catalog No. N270733PEC5


Add to cart

Description

Description

Fumarase Monoclonal antibody specifically detects Fumarase in Human, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Fumarase
OTI1F10
Flow Cytometry
Affinity Purified, Conjugated
FH
Full length recombinant protein of human Fumarase (NP_000134) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
2271.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit