Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fumonisin B2, 96+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Fumonisin B2 is a fumonisin mycotoxin produced by the fungus Fusarium
• Fumonisin B2 is a structural analog of fumonisin B1 but it is more cytotoxic than B1 and can inhibit sphinganine-N-acetyltransferase (ceramide synthase)
Applications
• In vitro and in vivo studies have shown Fumonisin B2 can induce apoptosis
• It can inhibit de novo sphingolipid biosynthesis
• It is also considered an Aspergillus metabolite
• It shows cytotoxicity against a panel of seven rat hepatoma cell lines, and MDCK cell line
• It induces hepatocyte nodule formation, a marker of cancer initiation, in rats
Specifications
Assay Percent Range | 96+% |
Quantity | 1 mg |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
Formula Weight | 759.9 |
Percent Purity | 96+% |
Chemical Name or Material | Fumonisin B2 |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction