Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Funaltrexamine hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

72786-10-8
C25H31ClN2O6
490.981
BIPHUOBUKMPSQR-RMEGLGEHSA-N
(E)-4-[[5α,6β)-17-Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]amino]-4-oxo-2-butenoicacid methyl ester hydrochloride
124081002

Irreversible μ-selective antagonist

Selective μ opioid receptor antagonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SMILES COC(=O)C=CC(=O)NC1CCC2(C3CC4=C5C2(C1OC5=C(C=C4)O)CCN3CC6CC6)O.Cl
IUPAC Name methyl (E)-4-[[(4R,4aR,7R,7aR,12bR)-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-yl]amino]-4-oxobut-2-enoate;hydrochloride
Formula Weight 490.99
Quantity 10mg
Chemical Name or Material β-Funaltrexamine hydrochloride
Grade >98%

Recommended Storage : Desiccate at -20°C