Learn More
Function Generator Products for Science Education
Function generator assembly kit.
Manufacturer:  Elenco Electronics IncSupplier Diversity Partner ELENCO FG500K
Catalog No. S65259
Add to cart
Description
Description
  • Complete with all parts, PC board, schematic, deluxe case with handle and extensive assembly manual
  • Provides sine, square, or sawtooth waveforms continuously variable at 1Hz to 1MHz
  • Amplitude adjustable to 3.5V peak to peak
  • Requires soldering
Specifications
Specifications
1Hz to 1MHz
Complete function generator capable of producing sine, square and triangle wave forms, experimentation and applications by the student
FG-500
9 to 18VDC
PC board, schematic, deluxe case with handle and extensive assembly manual
9 to 18VDC 1Hz to 1MHz
SDS
Documents
Accessories