Fungilab ASTM D-1200 Flow Cup Viscometers

Manufacturer: Fungilab FCA002

 View more versions of this product

Catalog No. 06-815-181

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D1200, calibration certificate
    Includes Calibration certificate
    Size 2
    Viscosity 25 to 120 cSt
    For Use With (Application) Used in paint, ink, varnishes and similar products industries
    Product Type Flow Cup Viscometer
    ASTM Numbers 2
    Calibrated Yes
    Material Anodized aluminum body, stainless steel orifice

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit