Learn More
Fungilab ASTM D-1200 Flow Cup Viscometers
Manufacturer:  Fungilab FCA004
 View more versions of this product
Catalog No. 06-815-183
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
4
ASTM D1200, calibration certificate
Anodized aluminum body, stainless steel orifice
70 to 370 cSt
Flow Cup Viscometer
Yes
Calibration certificate
4
Used in paint, ink, varnishes and similar products industries