Fungilab ASTM D-1200 Flow Cup Viscometers

Manufacturer: Fungilab FCA003

 View more versions of this product

Catalog No. 06-815-182

  Check Availability
Add to Cart

    Specifications

    ASTM Numbers 3
    Calibrated Yes
    Certifications/Compliance ASTM D1200, calibration certificate
    Includes Calibration certificate
    Material Anodized aluminum body, stainless steel orifice
    Size 3
    For Use With (Application) Used in paint, ink, varnishes and similar products industries
    Product Type Flow Cup Viscometer
    Viscosity 40 to 220 cSt

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit