Learn More
Fungilab BS/IP/RF U-Tube Reverse Flow Capillary Viscometers
Manufacturer:  Fungilab CV006004
 View more versions of this product
Catalog No. 06-815-169
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
4
BS, ASTM D 445, D446, ISO 3104, ISO 3105 and UNE 400313, calibration certificate
Borosilicate Glass
11mL
4
0.55 lb.
For opaque liquids
Yes
Calibration certificate
±0.2%
0.37 oz.
20 to 100 cSt
0.25kg
Bs/Ip/Rf U-Tube Reverse Flow Viscometer