Learn More
Fungilab BS/U-Tube Capillary Viscometers
Manufacturer:  Fungilab CV006108
 View more versions of this product
Catalog No. 06-815-165
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
H
BS, UNE 400313, ASTM D 445, D446, ISO 3104, ISO 3105 , calibration certificate
Borosilicate Glass
11mL
H
0.55 lb.
For transparent liquids
Yes
Calibration certificate
±0.2%
0.37 oz.
2.000 to 10.000 cSt
0.25kg
Bs/U U-Tube Reverse Flow Viscometer