Learn More
Fungilab™ BS/U-Tube Capillary Viscometers
$123.00 - $174.20
Specifications
Calibrated Yes
Certifications/Compliance BS, UNE 400313, ASTM D 445, D446, ISO 3104, ISO 3105 , calibration certificate
For Use With (Application) For transparent liquids
Product Type Bs/U U-Tube Reverse Flow Viscometer
Includes Calibration certificate
View More Specs
Products 8
Catalog Number Mfr. No. ASTM Numbers Size Viscosity Price Quantity & Availability  
Catalog Number Mfr. No. ASTM Numbers Size Viscosity Price Quantity & Availability  
06-815-161 View Documents Fungilab™
CV006104
D D 20 to 100 cSt Each for $128.66
Add to cart
06-815-162 View Documents Fungilab™
CV006105
E E 60 to 300 cSt Each for $123.00
Add to cart
06-815-160 View Documents Fungilab™
CV006103
C C 6 to 30 cSt Each for $172.00
Add to cart
06-815-163 View Documents Fungilab™
CV006106
F F 200 to 1.000 cSt Each for $128.66
Add to cart
06-815-164 View Documents Fungilab™
CV006107
G G 600 to 3.000 cSt Each for $128.66
Add to cart
06-815-159 View Documents Fungilab™
CV006102
B B 2 to 10 cSt Each for $127.40
Add to cart
06-815-158 View Documents Fungilab™
CV006101
A A 0.9 to 3 cSt Each for $174.20
Add to cart
06-815-165 View Documents Fungilab™
CV006108
H H 2.000 to 10.000 cSt Each for $128.66
Add to cart
Specifications
Specifications
Yes
For transparent liquids
Calibration certificate
±0.2%
0.37 oz.
0.25kg
BS, UNE 400313, ASTM D 445, D446, ISO 3104, ISO 3105 , calibration certificate
Bs/U U-Tube Reverse Flow Viscometer
Borosilicate Glass
11mL
0.55 lb.