Learn More
Fungilab BS/U-Tube Capillary Viscometers
Manufacturer:  Fungilab CV006101
 View more versions of this product
Catalog No. 06-815-158
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
A
BS, UNE 400313, ASTM D 445, D446, ISO 3104, ISO 3105 , calibration certificate
Borosilicate Glass
11mL
A
0.55 lb.
For transparent liquids
Yes
Calibration certificate
±0.2%
0.37 oz.
0.9 to 3 cSt
0.25kg
Bs/U U-Tube Reverse Flow Viscometer