Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fungilab Cannon-Fenske Routine Viscometer

Manufacturer:  Fungilab CV004207

 View more versions of this product

Catalog No. 06-815-121


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

300
Yes
Calibration certificate
±0.2%
0.23 oz.
50 to 200 cSt
0.25kg
Cannon™ Fenske Routine Viscometer
0.25
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
Glass
7mL
300
0.55 lb.
Measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit