Learn More
Fungilab™ Cannon-Fenske Routine Viscometer
$135.00 - $137.60
Specifications
Calibrated Yes
Certifications/Compliance ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
For Use With (Application) Measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries
Product Type Cannon™ Fenske Routine Viscometer
Includes Calibration certificate
View More Specs
Products 9
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity & Availability  
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity & Availability  
06-815-119 View Documents Fungilab™
CV004205
0.035 150 150 7 to 35 cSt Each for $136.00
Add to cart
06-815-116 View Documents Fungilab™
CV004202
0.004 50 50 0.8 to 4 cSt Each for $136.00
Add to cart
06-815-118 View Documents Fungilab™
CV004204
0.015 100 100 3 to 15 cSt Each for $137.60
Add to cart
06-815-120 View Documents Fungilab™
CV004206
0.1 200 200 20 to 100 cSt Each for $136.00
Add to cart
06-815-121 View Documents Fungilab™
CV004207
0.25 300 300 50 to 200 cSt Each for $136.00
Add to cart
06-815-123 View Documents Fungilab™
CV004209
1.2 400 400 240 to 1.200 cSt Each for $136.00
Add to cart
06-815-125 View Documents Fungilab™
CV004211
8 500 500 1.600 to 8.000 cSt Each for $135.00
Add to cart
06-815-124 View Documents Fungilab™
CV004210
2.5 450 450 500 to 2500 cSt Each for $135.00
Add to cart
06-815-126 View Documents Fungilab™
CV004212
20 600 600 4.000 to 20.000 cSt Each for $135.00
Add to cart
Specifications
Specifications
Yes
Measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries
Calibration certificate
±0.2%
0.23 oz.
0.25kg
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
Cannon™ Fenske Routine Viscometer
Glass
7mL
0.55 lb.