Learn More
Fungilab Eagle Series Thermostatic Water Baths
Manufacturer:  Fungilab B150
 View more versions of this product
Catalog No. 06-815-195
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5° to 150°C
12.9 in.
EAGLE 150
5.1 in.
0.3°C
FUNGILAB viscometer and accessories
9.03 lb.
±0.03K
190 to 230V
33cm
1 to 2kW
190 to 230V, 50/60Hz
LED
Designed to fit perfectly all FUNGILAB viscometer and accessories
4.1kg