Learn More
Fungilab Glass Thermometers for Hoppler Viscometer
Glass thermometers for a variety of temperature ranges.
Manufacturer:  Fungilab V91004
 View more versions of this product
Catalog No. 06-815-101
  Check Availability
Add to cart
Specifications
Description
  • Can be used with Fungilab Visco Ball Viscometer
  • Divisions: 0.1°C
Description & Specifications
Specifications
DIN 53015, ISO 12058
165° to 214°F
Hoppler viscometer
Glass
74° to 101°C
Thermometer