Learn More
Fungilab ISO 2431 Flow Cup Viscometers
Manufacturer:  Fungilab FCI005
 View more versions of this product
Catalog No. 06-815-179
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5
ISO 2431, calibration certificate
Anodized aluminum body, stainless steel orifice
100 to 350 cSt
0.7kg
Flow Cup Viscometer
Yes
Calibration certificate
5
1.54 lb.
Used in paint, ink, varnishes and similar products industries