Fungilab ISO 2431 Flow Cup Viscometers

Manufacturer: Fungilab FCI006

 View more versions of this product

Catalog No. 06-815-180

  Check Availability
Add to Cart

    Specifications

    Calibrated Yes
    Viscosity 190 to 680 cSt
    Weight (English) 1.54 lb.
    ASTM Numbers 6
    Certifications/Compliance ISO 2431, calibration certificate
    Includes Calibration certificate
    Material Anodized aluminum body, stainless steel orifice
    Size 6
    Weight (Metric) 0.7kg
    For Use With (Application) Used in paint, ink, varnishes and similar products industries
    Product Type Flow Cup Viscometer

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit