Learn More
Fungilab Silicone Standard Oils
Silicone standard oils to check and calibrate rotational viscometers
Manufacturer:  Fungilab V12Z30
 View more versions of this product
Catalog No. 06-815-072
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
BS EN ISO / IEC 17025
Rotational viscometer
Each
20.28 oz.
2kg
600mL
Used for viscosity tests and meter calibration
Ready-to-use Packs
1,019 MPa at 25°C
4.4 lb.
Silicone Standard Oil
Safety and Handling
ShelfLife : 2 Years