Learn More
Fungilab Spindles for Small Sample Adapters
Stainless steel disc measuring system for relative viscosity measurements in a 600mL beaker.
Manufacturer:  Fungilab V12709N
 View more versions of this product
Catalog No. 06-815-056
  Check Availability
Add to cart
Specifications
Description
Spindles for use with Small Sample Adapters (Cat. Nos. 06-815-032, -031).
  • This system is used for measurements in accordance with the ASTM-ISO 2555 standard
  • Standard spindles (included in standard delivery): R and H spindles, L spindles
  • Special spindles (additionally ordered with small sample adapters APM and APM/B): TL and TR spindles
  • Spindle set L: 20 to 6,000,000 cP (for ADV and PRO models); 20 to 2,000,000 cP (for ONE model)
  • Spindle set R: 100 to 40,000,000cP
  • Spindles set R for H models: 200 to 106,000,000cP
Description & Specifications
Specifications
ASTM, ISO 2555
Stainless Steel
Small sample adapter
For relative viscosity measurement
R: 250 to 830,000/H: 20,000 to 6,700,000 cP
Tr9 Spindle