Learn More
Fungilab Support Stands for Flow Cup Viscometers
For use with Flow Cup Viscometers.
Manufacturer:  Fungilab FCS
 View more versions of this product
Catalog No. 06-815-184
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Precision level and an overflow glass draw plate
Specifications
Description
  • Tripod support
  • Adjustable
Description & Specifications
Specifications
ISO 2431, ASTM D 1200
Flow cup viscometer
Tripod Support