Learn More
Fungilab™ Ubbelohde ASTM and DIN Capillary Viscometers
$135.00 - $179.70
Specifications
Calibrated Yes
For Use With (Application) Used to determine kinematic viscosity of transparent Newtonian liquids
Includes Calibration certificate
Material Glass
Precision ±0.2%
View More Specs
Products 25
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Certifications/Compliance Size Price Quantity & Availability  
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Certifications/Compliance Size Price Quantity & Availability  
06-815-128 View Documents Fungilab™
CV003002
0.003 0C ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 0C Each for $179.70
Add to cart
06-815-127 View Documents Fungilab™
CV003001
0.001 0 ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 0 Each for $167.50
Add to cart
06-815-138 View Documents Fungilab™
CV003012
5 3B ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 3B Each for $165.00
Add to cart
06-815-134 View Documents Fungilab™
CV003008
0.3 2C ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 2C Each for $165.00
Add to cart
06-815-136 View Documents Fungilab™
CV003010
1 3 ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 3 Each for $167.50
Add to cart
06-815-139 View Documents Fungilab™
CV003013
10 4 ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 4 Each for $165.00
Add to cart
06-815-135 View Documents Fungilab™
CV003009
0.5 2B ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 2B Each for $165.00
Add to cart
06-815-140 View Documents Fungilab™
CV003014
30 4C ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 4C Each for $167.50
Add to cart
06-815-141 View Documents Fungilab™
CV003015
50 4B ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 4B Each for $165.00
Add to cart
06-815-142 View Documents Fungilab™
CV003016
100 5 ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate 5 Each for $165.00
Add to cart
06-815-144 View Documents Fungilab™
CV003101
0.003 0C DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate 0C Each for $165.00
Add to cart
06-815-145 View Documents Fungilab™
CV003102
0.005 0A DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate 0A Each for $167.50
Add to cart
06-815-143 View Documents Fungilab™
CV003100
0.001 0 DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate 0 Each for $135.00
Add to cart
06-815-146 View Documents Fungilab™
CV003103
0.01 I DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate I Each for $179.70
Add to cart
06-815-147 View Documents Fungilab™
CV003104
0.03 Ic DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate Ic Each for $165.00
Add to cart
06-815-148 View Documents Fungilab™
CV003111
0.05 Ia DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate Ia Each for $165.00
Add to cart
06-815-149 View Documents Fungilab™
CV003105
0.1 II DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate II Each for $167.50
Add to cart
06-815-150 View Documents Fungilab™
CV003106
0.3 IIc DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IIc Each for $165.00
Add to cart
06-815-154 View Documents Fungilab™
CV003113
5 IIIa DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IIIa Each for $165.00
Add to cart
06-815-151 View Documents Fungilab™
CV003112
0.5 IIa DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IIa Each for $165.00
Add to cart
06-815-152 View Documents Fungilab™
CV003107
1 III DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate III Each for $165.00
Add to cart
06-815-153 View Documents Fungilab™
CV003108
3 IIIc DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IIIc Each for $178.00
Add to cart
06-815-155 View Documents Fungilab™
CV003109
10 IV DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IV Each for $157.28
Add to cart
06-815-156 View Documents Fungilab™
CV003110
30 IVc DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IVc Each for $165.00
Add to cart
06-815-157 View Documents Fungilab™
CV003114
50 IVa DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate IVa Each for $165.00
Add to cart
Specifications
Specifications
Yes
Calibration certificate
±0.2%
0.5 oz.
0.25kg
Used to determine kinematic viscosity of transparent Newtonian liquids
Glass
15mL
0.55 lb.