Learn More
Fungilab™ Ubbelohde Capillary Viscometer
$135.00 - $180.10
Specifications
Calibrated Yes
Certifications/Compliance ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
For Use With (Application) Used to determine kinematic viscosity of transparent Newtonian liquids
Product Type Ubbelohde Astm Viscometer
Includes Calibration certificate
View More Specs
Products 6
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity & Availability  
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity & Availability  
06-815-129 View Documents Fungilab™
CV003003
0.01 0B 0B 1 to 5 cSt Each for $180.10
Add to cart
06-815-130 View Documents Fungilab™
CV003004
0.01 1 1 2 to 10 cSt Each for $180.10
Add to cart
06-815-131 View Documents Fungilab™
CV003005
0.03 1C 1C 6 to 30 cSt Each for $180.10
Add to cart
06-815-132 View Documents Fungilab™
CV003006
0.05 1B 1B 10 to 50 cSt Each for $167.00
Add to cart
06-815-133 View Documents Fungilab™
CV003007
0.1 2 2 20 to 100 cSt Each for $180.10
Add to cart
06-815-137 View Documents Fungilab™
CV003011
3 3C 3C 600 to 3.000 cSt Each for $135.00
Add to cart
Specifications
Specifications
Yes
Used to determine kinematic viscosity of transparent Newtonian liquids
Calibration certificate
±0.2%
0.5 oz.
0.25kg
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
Ubbelohde Astm Viscometer
Glass
15mL
0.55 lb.