Learn More
Fungilab™ Viscolead One Viscometer
Measure relative and absolute viscosity.
$2529.00 - $3070.00
Specifications
Alarm Types Auto-test with sound and visual malfunction alarm
Certifications/Compliance BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
Repeatability 0.2%
Hertz 50/60Hz
Speed Range 0.3 to 100rpm
View More Specs
Includes: Standard spindles (4 spindles for L model and 6 spindles for R and H models), viscometer stand and spindle protector
Products 6
Catalog Number Mfr. No. Product Type Viscosity Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Viscosity Includes Price Quantity & Availability  
06-815-017 View Documents Fungilab™
VL101002
Viscolead One R Rotational Viscometer 100 to 13.000.000 cP PPR spindle coupling system, viscometer stand and spindle protector, 6 standard spindles Each for $3,070.00
Add to cart
06-815-018 View Documents Fungilab™
VL101003
Viscolead One L Rotational Viscometer 20 to 2.000.000 cP PPR spindle coupling system, viscometer stand and spindle protector, 4 standard spindles Each for $2,839.00
Add to cart
06-815-016 View Documents Fungilab™
VL101001
Viscolead One H Rotational Viscometer 200 to 106,000,000 cP PPR spindle coupling system, viscometer stand and spindle protector, 6 standard spindles Each for $2,839.00
Add to cart
06-815-003 View Documents Fungilab™
VL100003
Viscolead One L Rotational Viscometer 20 to 2.000.000 cP Viscometer stand and spindle protector, 4 standard spindles Each for $2,570.00
Add to cart
06-815-002 View Documents Fungilab™
VL100002
Viscolead One R Rotational Viscometer 100 to 13.000.000 cP Viscometer stand and spindle protector, 6 standard spindles Each for $2,570.00
Add to cart
06-815-001 View Documents Fungilab™
VL100001
Viscolead One H Rotational Viscometer 200 to 106,000,000 cP Viscometer stand and spindle protector, 6 standard spindles Each for $2,529.00
Add to cart
Description
Description
Digital display of selected speed (rpm), selected spindle (SP), viscosity reading in (cP/mPaS) and Percentage of full scale (%).
  • SI to CGS Unit converter
  • AUTO-TEST with sound and visual malfunction alarm
  • AUTO-RANGE function
  • User-enabled calibration
  • Option of 10 languages
  • AISI 316 stainless steel spindles, identifiable by number and letter
Order Info
Also available with the Push Plug and Release (PPR) System for spindle coupling.
Compliance
Conforms to standards:
BS: 6075, 5350
ISO: 2555, 1652
ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
Specifications
Specifications
Auto-test with sound and visual malfunction alarm
0.2%
0.3 to 100rpm
BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
50/60Hz
100/240VAC