Learn More
Sklar InstrumentsSupplier Diversity Partner Funnel, Sklar, Stainless Steel. This product is a 6-1/4 in. x 6-5/8 in. funnel with a 22-1/4 oz capacity. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101160
Funnel, Sklar, Stainless Steel. Sklar's Surgical Funnel features a hanging hook so it can easily be elevated above a patient's head when introduction of fluid is necessary as part of a surgical procedure. This product is a 6-1/4 in. x 6-5/8 in. funnel with a 22-1/4 oz capacity.
Catalog No. 50-143-6933