Learn More
Synthware Filter Funnels with Perforated Glass Plate
Used in conjunction with vacuum source to filter and collect fluids
Manufacturer: Synthware F932450
Description
- 24/40 standard taper inner joint
- Hose connector for connection to vacuum source
- Glass plate O.D.: 24mm
Specifications
24 mm Plate | |
Filter Funnel with perforated glass plate | |
50 mm |