Tocris™ Furamidine dihydrochloride

Selective PRMT1 inhibitor

Manufacturer: Tocris™ 5202/10

 View more versions of this product

Catalog No. 52-021-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Description Furamidine dihydrochloride
    Quantity 10mg
    Product Type Furamidine dihydrochloride
    Molecular Weight 377.27
    Purity >98

    Selective protein arginine methyltransferase 1 (PRMT1) inhibitor (IC50 = 9.4µM). Exhibits selectivity over PRMT5, PRMT6 and CARM1 (IC50 values are 166, 283 and >400µM respectively). Inhibits cell proliferation in leukemia cell lines. Cell-permeable.

    • Chemical Name: 4,4'-(2,5-Furandiyl)bis-benzenecarboximidamide dihydrochloride
    • Formula: C18H16N4O.2HCl
    • CAS Number: 55368-40-6
    • Solubility: Soluble to 50mM in water and to 50mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit