Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furan-2-acetic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
2745-26-8
C6H6O3
126.11
VYSRZETUSAOIMP-UHFFFAOYSA-N
2-furan-2-yl acetic acid,2-furanacetic acid,2-furylacetic acid,furan-2-acetic acid,furylacetic acid,unii-c2xfe1jto5,2-furylaceticacid,furan-2-yl-acetic acid,c2xfe1jto5,2-2-furyl acetic acid
75974
2-(furan-2-yl)acetic acid
C1=COC(=C1)CC(=O)O
2745-26-8
C6H6O3
126.11
VYSRZETUSAOIMP-UHFFFAOYSA-N
2-furan-2-yl acetic acid,2-furanacetic acid,2-furylacetic acid,furan-2-acetic acid,furylacetic acid,unii-c2xfe1jto5,2-furylaceticacid,furan-2-yl-acetic acid,c2xfe1jto5,2-2-furyl acetic acid
75974
2-(furan-2-yl)acetic acid
C1=COC(=C1)CC(=O)O
Packaging Glass Bottle
Melting Point 64.0°C to 69.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Furan-2-acetic acid

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 220-380-3