Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Furan-2-boronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
13331-23-2
C4H5BO3
111.891
MFCD00799544
PZJSZBJLOWMDRG-UHFFFAOYSA-N
2-furanboronic acid,furan-2-boronic acid,2-furylboronic acid,2-furanylboronic acid,furan-2-ylboranediol,furan 2-boronic acid,2-furan boronic acid,furan-2-yl boronic acid,boronic acid, furanyl,2furylboronic acid
2734357
furan-2-ylboronic acid
B(C1=CC=CO1)(O)O
13331-23-2
C4H5BO3
111.891
MFCD00799544
PZJSZBJLOWMDRG-UHFFFAOYSA-N
2-furanboronic acid,furan-2-boronic acid,2-furylboronic acid,2-furanylboronic acid,furan-2-ylboranediol,furan 2-boronic acid,2-furan boronic acid,furan-2-yl boronic acid,boronic acid, furanyl,2furylboronic acid
2734357
furan-2-ylboronic acid
B(C1=CC=CO1)(O)O
Applications
It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 5g
Melting Point ~120°C (decomposition)
Beilstein 112370
Formula Weight 111.89
Percent Purity 97%
Chemical Name or Material Furan-2-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C