Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furan-2-boronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
13331-23-2
C4H5BO3
111.89
MFCD00799544
PZJSZBJLOWMDRG-UHFFFAOYSA-N
2-furanboronic acid,furan-2-boronic acid,2-furylboronic acid,2-furanylboronic acid,furan-2-ylboranediol,furan 2-boronic acid,2-furan boronic acid,furan-2-yl boronic acid,boronic acid, furanyl,2furylboronic acid
2734357
furan-2-ylboronic acid
B(C1=CC=CO1)(O)O
13331-23-2
C4H5BO3
111.89
MFCD00799544
PZJSZBJLOWMDRG-UHFFFAOYSA-N
2-furanboronic acid,furan-2-boronic acid,2-furylboronic acid,2-furanylboronic acid,furan-2-ylboranediol,furan 2-boronic acid,2-furan boronic acid,furan-2-yl boronic acid,boronic acid, furanyl,2furylboronic acid
2734357
furan-2-ylboronic acid
B(C1=CC=CO1)(O)O
Quantity 10g
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Formula Weight 111.89
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Furan-2-boronic acid

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning